missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) nitrate, ACS, 99.0% min, Thermo Scientific Chemicals

Product Code. 11307817
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

For Use As An Oxidizer in Dye Industry, in Manufacturing of Mother-of-Pearl, As Mordant in Printing Textile

As an oxidizer in dye industry, in manufacturing of mother-of-pearl, as mordant in printing textile, as sensitizer in photographyEmployed in industrial applications such as heat stabilization in nylon and polyesters, coatings of photothermographic paper, gold cyanidation and pyrotechnics. Finds

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10099-74-8
Molecular Formula N2O6Pb
Molecular Weight (g/mol) 331.20
MDL Number MFCD00011153
InChI Key RLJMLMKIBZAXJO-UHFFFAOYSA-N
Synonym lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
PubChem CID 24924
ChEBI CHEBI:37187
SMILES [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 470°C (decomposition)
Color White
Physical Form Crystals
Quantity 500 g
Assay Percent Range 99.0% min.
Solubility Information Freely soluble in water. Soluble in absolute alcohol,methanol. Insoluble in concentrated HNO3
IUPAC Name lead(2+);dinitrate
Formula Weight 331.2
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline
Vapor Pressure approx.3.0-4.0 (20% aqueous solution)
Density 4.53 g/cm3 at 20°C
Chemical Name or Material Lead(II) nitrate, For ACS analysis
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) nitrate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P201-Obtain special instructions before use.
  • P220-Keep/Store away from clothing/combustible materials.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'