missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) nitrate, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 470.0°C |
Color | White |
Physical Form | Crystalline Powder |
Quantity | 5 g |
Linear Formula | Pb(NO3)2 |
Merck Index | 15, 5469 |
Solubility Information | Solubility in water: 343g/L. |
IUPAC Name | λ²-lead(2+) dinitrate |
Formula Weight | 331.2 |
Percent Purity | ≥99.0% |
Show More |
Safety and Handling
Product Identifier
- Lead(II) nitrate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 2
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'