missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) nitrate, Puriss., ≥99%, Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
50 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 1 kg |
Linear Formula | Pb(NO3)2 |
UN Number | 1469 |
SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Pb+2] |
IUPAC Name | lead(2+);dinitrate |
Formula Weight | 331.21g/mol |
Percent Purity | ≥99% |
Grade | puriss. |
Chemical Name or Material | Lead(Ii) Nitrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'