missing translation for 'onlineSavingsMsg'
Learn More
Lead (II) Nitrate solution 0.5mol/L, Fisher Chemical™ Product Code.: 12953554
compliance-icons compliance-icons compliance-icons

Lead (II) Nitrate solution 0.5mol/L, Fisher Chemical™

Product Code. 12953554
1000 mL, Bottle
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
This item is not returnable. View return policy

Product Code. 12953554

Brand: Fisher Chemical VL 238 1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

CAS 10099-74-8
Quantity 1000 mL
Molecular Formula N2O6Pb
MDL Number MFCD00011153
UN Number 3082
Synonym lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
InChI Key RLJMLMKIBZAXJO-UHFFFAOYSA-N
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Pb+2]
IUPAC Name lead(2+);dinitrate
Molecular Weight (g/mol) 331.20
PubChem CID 24924
ChEBI CHEBI:37187
Concentration or Composition (by Analyte or Components) 0.5M (1N)
Packaging Bottle
Chemical Name or Material Lead Nitrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead nitrate 0.5M Volumetric Solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360-May damage fertility or the unborn child.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead (II) Nitrate solution 0.5mol/L, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.