missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) oxide, 99+%, Thermo Scientific Chemicals

Product Code. 10481241
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-36-8
Molecular Formula OPb
Molecular Weight (g/mol) 223.20
MDL Number MFCD00011164
InChI Key YEXPOXQUZXUXJW-UHFFFAOYSA-N
Synonym lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
PubChem CID 14827
IUPAC Name plumbanone
SMILES O=[Pb]

Specifications

Boiling Point 1470.0°C
Melting Point 886.0°C
Color Yellow
Physical Form Fine Powder
Quantity 1 kg
Assay Percent Range 99.6% min. (Complexometry)
Linear Formula PbO
Fieser 01,536; 02,233
Merck Index 15, 5468
Solubility Information Solubility in water: 70.2 mg/l (20°C).
Formula Weight 223.19
Percent Purity 99.7%
Packaging Glass bottle
Chemical Name or Material Lead(II) oxide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity Category 1
  • Carcinogenicity Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Reproductive toxicity on or via lactation
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H351-Suspected of causing cancer.
  • H372-Causes damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H362-May cause harm to breast-fed children.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash hands and any exposed skin thoroughly after handling.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'