Learn More
Lead(II) oxide, 99+%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 1317-36-8 |
---|---|
Molecular Formula | OPb |
Molecular Weight (g/mol) | 223.20 |
MDL Number | MFCD00011164 |
InChI Key | YEXPOXQUZXUXJW-UHFFFAOYSA-N |
Synonym | lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow |
PubChem CID | 14827 |
IUPAC Name | plumbanone |
SMILES | O=[Pb] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1317-36-8 | |
223.20 | |
YEXPOXQUZXUXJW-UHFFFAOYSA-N | |
14827 | |
O=[Pb] |
OPb | |
MFCD00011164 | |
lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow | |
plumbanone |
Specifications
1470.0°C | |
1317-36-8 | |
Fine Powder | |
OPb | |
MFCD00011164 | |
15, 5468 | |
Solubility in water: 70.2 mg/l (20°C). | |
O=[Pb] | |
223.20 | |
223.19 | |
Lead(II) oxide |
886.0°C | |
Yellow | |
99.6% min. (Complexometry) | |
PbO | |
01,536; 02,233 | |
lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow | |
YEXPOXQUZXUXJW-UHFFFAOYSA-N | |
plumbanone | |
14827 | |
99.7% |
Safety and Handling
GHS H Statement
May damage the unborn child.
Suspected of damaging fertility.
Causes damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Harmful if swallowed.
Suspected of causing cancer.
GHS P Statement
Use personal protective equipment as required.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON C
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 215-267-
- Lead(II) oxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity Category 1
- Carcinogenicity Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- Reproductive toxicity on or via lactation
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H351-Suspected of causing cancer.
- H372-Causes damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- H362-May cause harm to breast-fed children.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash hands and any exposed skin thoroughly after handling.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
RUO – Research Use Only