missing translation for 'onlineSavingsMsg'
Learn More
Lead(II) Oxide, Extra Pure, SLR, Fisher Chemical Product Code.: 10090230

Lead(II) Oxide, Extra Pure, SLR, Fisher Chemical

Product Code. 10090230
500 g, Plastic container
Click to view available options
Quantity:
500 g
Packaging:
Plastic container
This item is not returnable. View return policy

Product Code. 10090230

Brand: Fisher Chemical L/1600/53

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 1470°C
Melting Point 886°C
CAS 1317-36-8
Color Yellow
Physical Form Solid
Quantity 500 g
Molecular Formula OPb
MDL Number MFCD00011164
Synonym lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
InChI Key YEXPOXQUZXUXJW-UHFFFAOYSA-N
SMILES O=[Pb]
IUPAC Name plumbanone
Molecular Weight (g/mol) 223.20
PubChem CID 14827
Formula Weight 223.19g/mol
Packaging Plastic container
Vapor Pressure 10mmHg 1085
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H351-Suspected of causing cancer.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyA§ thoroughly after handling.
  • P281-Use personal protective equipment as required.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/A§/if you feel unwell.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead(II) Oxide, Extra Pure, SLR, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.