missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) oxide, low silver, Thermo Scientific Chemicals

Product Code. 11347658
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Lead(II) oxide is used in storage batteries, ceramic cements and fluxes, pottery and glazes, in chromium pigments, in oil refining, in paints, and in the assay of precious metal ores.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • As storage batteries, ceramic cements and fluxes, pottery and glazes, in chromium pigments, in oil refining, in paints, and in the assay of precious metal ores
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-36-8
Molecular Formula OPb
Molecular Weight (g/mol) 223.20
MDL Number MFCD00011164
InChI Key YEXPOXQUZXUXJW-UHFFFAOYSA-N
Synonym lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
PubChem CID 14827
SMILES O=[Pb]

Specifications

Melting Point 886°C
Physical Form Powder
Quantity 2 kg
UN Number UN3077
Merck Index 14,5413
Solubility Information Insoluble in water and alcohol; Soluble in acetic acid,dilute HNO3 and alkalies
IUPAC Name oxolead
Formula Weight 223.19
Odor Odorless
Density 9.53 g/mL
Chemical Name or Material Lead(II) oxide, Low silver
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'