missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11365047
Click to view available options
Quantity:
25 g
100 g
500 g

Used in producing iridescent colors on brass and bronze, coloring sulfur-containing substances and used as pigment for rubber. It is also used in oil refining, varnishes, paints, enamels and in assays for precious metal ores.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-36-8
Molecular Formula OPb
Molecular Weight (g/mol) 223.20
MDL Number MFCD00011164
InChI Key YEXPOXQUZXUXJW-UHFFFAOYSA-N
Synonym lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
PubChem CID 14827
IUPAC Name oxolead
SMILES O=[Pb]

Specifications

Melting Point 886°C
Color Yellow
Physical Form Powder
Quantity 100 g
UN Number UN3077
Merck Index 14,5413
Solubility Information Insoluble in water,soluble in concentrated alkalies,hydrochloric acid and ammonium chloride.
Formula Weight 223.19
Percent Purity 99.999%
Grade Puratronic™
Odor Odorless
Density 9.53 g/mL
Assay (metals basis)
Chemical Name or Material Lead(II) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.