missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) oxide, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals

99.09 € - 387.93 €

Chemical Identifiers

CAS 1317-36-8
Molecular Formula OPb
Molecular Weight (g/mol) 223.20
MDL Number MFCD00011164
InChI Key YEXPOXQUZXUXJW-UHFFFAOYSA-N
Synonym lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
PubChem CID 14827
SMILES O=[Pb]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11306807
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010728.14
25 g
99.09 €
25g
Please to purchase this item. Need a web account? Register with us today!
11316807
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010728.22
100 g
387.93 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used in the manufacture of glass, ceramics and computer components. Addition of lead oxide increases the refractive index, electrical resistivity, Curie temperature, and the ability of the glass to absorb X-rays. It is also added to decrease the viscosity of glass. It is employed in the vulcanization of rubber, lead acid batteries, and in the production of certain pigments and paints. In cathode ray tube glass lead oxide is used to block X-ray emission. It acts as an efficient reagent in oxidation reactions as well as condensation reactions of hydroxy ketones with diamines to form quinoxaline derivatives with high yield.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1317-36-8
223.20
YEXPOXQUZXUXJW-UHFFFAOYSA-N
14827
OPb
MFCD00011164
lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
O=[Pb]

Specifications

886°C
Yellow
OPb
MFCD00011164
14,5413
Soluble in concentrated alkali,HCl and ammonium chloride. Insoluble in dilute alkali and alcohol.
O=[Pb]
223.20
223.19
Puratronic™
9.53 g/mL
Lead(II) oxide
1317-36-8
Powder
PbO
UN3077
lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
YEXPOXQUZXUXJW-UHFFFAOYSA-N
plumbanone
14827
99.9995%
Odorless
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c

H302+H332-H350-H360Df-H372

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

missing translation for 'einecsNumber' : 215-267-0

missing translation for 'rtecsNumber' : OG1750000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Lead(II) oxide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only