Learn More
Lead(II) perchlorate trihydrate, 99%, Thermo Scientific Chemicals
36.38 € - 553.76 €
Chemical Identifiers
CAS | 13453-62-8 |
---|---|
Molecular Formula | Cl2H6O11Pb |
Molecular Weight (g/mol) | 460.10 |
MDL Number | MFCD00149817 |
InChI Key | KKGGEAQRICYXNM-UHFFFAOYSA-L |
Synonym | lead ii perchlorate trihydrate, acmc-20aknd, 2clo4.pb.3h2o, lead perchlorate 3h2o, lead ii perchlorate, trihydrate, lead perchlorate, trihydrate, reagent acs, lead ii perchlorate trihydrate, acs reagent |
PubChem CID | 25021834 |
SMILES | O.O.O.[Pb++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10563021
|
Thermo Scientific Acros
254590250 |
25 g | Glass bottle |
36.38 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10378420
|
Thermo Scientific Acros
254592500 |
250 g | Glass bottle |
182.91 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10137463
|
Thermo Scientific Acros
254590010 |
1 kg | Glass bottle |
553.76 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
13453-62-8 | |
460.10 | |
KKGGEAQRICYXNM-UHFFFAOYSA-L | |
25021834 |
Cl2H6O11Pb | |
MFCD00149817 | |
lead ii perchlorate trihydrate, acmc-20aknd, 2clo4.pb.3h2o, lead perchlorate 3h2o, lead ii perchlorate, trihydrate, lead perchlorate, trihydrate, reagent acs, lead ii perchlorate trihydrate, acs reagent | |
O.O.O.[Pb++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O |
Specifications
100.0°C | |
White to Beige | |
Adhering Crystals | |
Cl2H6O11Pb | |
MFCD00149817 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol | |
O.O.O.[Pb++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O | |
460.10 | |
460.14 | |
2.6 | |
Lead(II) perchlorate trihydrate, 99% |
13453-62-8 | |
3 to 5 (5% soln.) | |
98.5% min. (Complexometry) | |
Pb(ClO4)2·3H2O | |
lead ii perchlorate trihydrate, acmc-20aknd, 2clo4.pb.3h2o, lead perchlorate 3h2o, lead ii perchlorate, trihydrate, lead perchlorate, trihydrate, reagent acs, lead ii perchlorate trihydrate, acs reagent | |
KKGGEAQRICYXNM-UHFFFAOYSA-L | |
lead(2+);diperchlorate;trihydrate | |
25021834 | |
99% | |
2.6000g/mL |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
May damage the unborn child. Suspected of damaging fertility.
May intensify fire; oxidizer.
GHS P Statement:
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- Lead(II) perchlorate trihydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Oxidising solid Category 2
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only