missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) perchlorate trihydrate, ACS, 97.0-102.0%, Thermo Scientific Chemicals

Product Code. 11341748
Click to view available options
Quantity:
100 g
500 g

Titrant in complexometric titrations

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13453-62-8
Molecular Formula Cl2H6O11Pb
Molecular Weight (g/mol) 460.10
MDL Number MFCD00149817
InChI Key KKGGEAQRICYXNM-UHFFFAOYSA-L
Synonym lead ii perchlorate trihydrate, acmc-20aknd, 2clo4.pb.3h2o, lead perchlorate 3h2o, lead ii perchlorate, trihydrate, lead perchlorate, trihydrate, reagent acs, lead ii perchlorate trihydrate, acs reagent
PubChem CID 25021834
IUPAC Name lead(2+);diperchlorate;trihydrate
SMILES O.O.O.[Pb++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Melting Point 100°C (decomposition)
Color White
Physical Form Crystalline
Quantity 100 g
Linear Formula Pb(ClO4)2·3H2O
UN Number UN1470
Solubility Information Soluble in water and alcohol.
Formula Weight 460.15 (406.10 Anhydrous)
Percent Purity ≤97%
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Density 2.60 g/mL
Chemical Name or Material Lead(II) perchlorate trihydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead (II) perchlorate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 1
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Indicaciones de peligro
  • H271-May cause fire or explosion; strong oxidiser.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Consejos de prudencia
  • P283-Wear fire/flame resistant/retardant clothing.
  • P306+P360-IF ON CLOTHING: rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado