missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) perchlorate trihydrate, ACS reagent, Thermo Scientific Chemicals

Product Code. 13555020
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13453-62-8
Molecular Formula Cl2H6O11Pb
Molecular Weight (g/mol) 460.10
MDL Number MFCD00149817
InChI Key KKGGEAQRICYXNM-UHFFFAOYSA-L
Synonym lead ii perchlorate trihydrate, acmc-20aknd, 2clo4.pb.3h2o, lead perchlorate 3h2o, lead ii perchlorate, trihydrate, lead perchlorate, trihydrate, reagent acs, lead ii perchlorate trihydrate, acs reagent
PubChem CID 25021834
IUPAC Name lead(2+);diperchlorate;trihydrate
SMILES O.O.O.[Pb++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Melting Point 100.0°C
Quantity 500 g
Linear Formula Pb(ClO4)2·3H2O
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 460.14
Grade ACS Reagent
Packaging Plastic bottle
Specific Gravity 2.6
Density 2.6000g/mL
Chemical Name or Material Lead(II) perchlorate trihydrate
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) perchlorate trihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Oxidising solid Category 2
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'