Learn More
Lead(II) sulfate, 99%, Thermo Scientific Chemicals
31.80 € - 80.80 €
Chemical Identifiers
CAS | 7446-14-2 |
---|---|
Molecular Formula | O4PbS |
Molecular Weight (g/mol) | 303.25 |
MDL Number | MFCD00011166 |
InChI Key | KEQXNNJHMWSZHK-UHFFFAOYSA-L |
Synonym | lead ii sulfate, lead sulfate, anglislite, lead sulphate, mulhouse white, lead bottoms, lead monosulfate, fast white, milk white, natural anglesite |
PubChem CID | 24008 |
IUPAC Name | lead(2+);sulfate |
SMILES | [O-]S(=O)(=O)[O-].[Pb+2] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10686923
|
Thermo Scientific Acros
387271000 |
100 g | Plastic bottle |
31.80 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10648614
|
Thermo Scientific Acros
387275000 |
500 g | Plastic bottle |
80.80 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
7446-14-2 | |
303.25 | |
KEQXNNJHMWSZHK-UHFFFAOYSA-L | |
24008 | |
[O-]S(=O)(=O)[O-].[Pb+2] |
O4PbS | |
MFCD00011166 | |
lead ii sulfate, lead sulfate, anglislite, lead sulphate, mulhouse white, lead bottoms, lead monosulfate, fast white, milk white, natural anglesite | |
lead(2+);sulfate |
Specifications
1170.0°C | |
White | |
99% | |
MFCD00011166 | |
lead ii sulfate, lead sulfate, anglislite, lead sulphate, mulhouse white, lead bottoms, lead monosulfate, fast white, milk white, natural anglesite | |
KEQXNNJHMWSZHK-UHFFFAOYSA-L | |
lead(2+);sulfate | |
24008 | |
99% | |
Lead(II) sulfate |
7446-14-2 | |
Crystalline Powder | |
O4PbS | |
15, 5475 | |
Solubility in water: slightly soluble | |
[O-]S(=O)(=O)[O-].[Pb+2] | |
303.25 | |
303.25 | |
Confirmed |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
May damage the unborn child.
Suspected
GHS P Statement
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a pos
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-198-9
- Lead(II) sulfate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only