Learn More
Lead(II) sulfide, 99.995% (metals basis), Thermo Scientific Chemicals
78.79 € - 293.46 €
Chemical Identifiers
CAS | 1314-87-0 |
---|---|
Molecular Formula | PbS |
Molecular Weight (g/mol) | 239.30 |
MDL Number | MFCD00016280 |
InChI Key | FILXVLQXZYXBSJ-UHFFFAOYSA-N |
Synonym | lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide |
PubChem CID | 14819 |
SMILES | [S--].[Pb++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15422287
|
Thermo Scientific Alfa Aesar
045462.04 |
2 g |
78.79 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15432287
|
Thermo Scientific Alfa Aesar
045462.09 |
10 g |
293.46 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lead(II) sulfide is used in infrared detectors, photo optic applications, slip property modifier used in friction industry for enhancing heat conduction and regulating friction coefficient.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1314-87-0 | |
239.30 | |
FILXVLQXZYXBSJ-UHFFFAOYSA-N | |
14819 |
PbS | |
MFCD00016280 | |
lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide | |
[S--].[Pb++] |
Specifications
1314-87-0 | |
Lump | |
PbS | |
lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide | |
[S--].[Pb++] | |
239.30 | |
239.25 | |
Black lumps | |
Lead(II) sulfide |
Black | |
99.995% (Metal basis) | |
MFCD00016280 | |
FILXVLQXZYXBSJ-UHFFFAOYSA-N | |
sulfanylidenelead | |
14819 | |
Odorless | |
7.5g/cm3 at 20°C |
Safety and Handling
H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.
missing translation for 'dotInformation' : DOT Class: 9, Packaging Group: III
missing translation for 'einecsNumber' : 215-246-6
- Lead(II) sulfide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.