Learn More
Lead(II) tantalum oxide, 99.9% (metals basis), Thermo Scientific™
85.00 € - 278.00 €
Chemical Identifiers
CAS | 12065-68-8 |
---|---|
Molecular Formula | O6PbTa2 |
Molecular Weight (g/mol) | 665.09 |
MDL Number | MFCD00054256 |
InChI Key | KNSIKTVGKTWOGA-UHFFFAOYSA-N |
Synonym | lead ii tantalum oxide, lead tantalate, lead ii tantalate |
PubChem CID | 16685095 |
IUPAC Name | (dioxotantaliooxy-$l^{2}-plumbanyl)oxy-dioxotantalum |
SMILES | O=[Ta](=O)O[Pb]O[Ta](=O)=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
12065-68-8 | |
665.09 | |
KNSIKTVGKTWOGA-UHFFFAOYSA-N | |
16685095 | |
O=[Ta](=O)O[Pb]O[Ta](=O)=O |
O6PbTa2 | |
MFCD00054256 | |
lead ii tantalum oxide, lead tantalate, lead ii tantalate | |
(dioxotantaliooxy-$l^{2}-plumbanyl)oxy-dioxotantalum |
Specifications
>350°C | |
Powder | |
MFCD00054256 | |
lead ii tantalum oxide, lead tantalate, lead ii tantalate | |
O=[Ta](=O)O[Pb]O[Ta](=O)=O | |
665.09 | |
665.09 | |
7.9 g/mL | |
Lead(II) tantalum oxide |
12065-68-8 | |
O6PbTa2 | |
UN2291 | |
KNSIKTVGKTWOGA-UHFFFAOYSA-N | |
(dioxotantaliooxy-$l^{2}-plumbanyl)oxy-dioxotantalum | |
16685095 | |
99.9% | |
(metals basis) |
Safety and Handling
GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H302+H332-H350-H360Df-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
missing translation for 'einecsNumber' : 235-065-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Lead(II) tantalum oxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Your input is important to us. Please complete this form to provide feedback related to the content on this product.