Learn More
Lead(II) telluride, 99.999% (metals basis), Thermo Scientific Chemicals
176.88 € - 593.96 €
Chemical Identifiers
CAS | 1314-91-6 |
---|---|
Molecular Formula | PbTe |
Molecular Weight (g/mol) | 334.80 |
MDL Number | MFCD00016274 |
InChI Key | OCGWQDWYSQAFTO-UHFFFAOYSA-N |
Synonym | lead telluride, lead ii telluride, unii-v1og6oa4bj, v1og6oa4bj, plumbanetellone, bleitellurid, lead monotelluride, lead ii telluride trace metals basis |
PubChem CID | 4389803 |
IUPAC Name | tellanylidenelead |
SMILES | [Te]=[Pb] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11319239
|
Thermo Scientific Alfa Aesar
088285.14 |
25 g |
176.88 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11329239
|
Thermo Scientific Alfa Aesar
088285.22 |
100 g |
593.96 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lead(II) telluride is used to make thermoelectric converter (cooler, generator, heat), infrared detectors and tunnel diode. It is also an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1314-91-6 | |
334.80 | |
OCGWQDWYSQAFTO-UHFFFAOYSA-N | |
4389803 | |
[Te]=[Pb] |
PbTe | |
MFCD00016274 | |
lead telluride, lead ii telluride, unii-v1og6oa4bj, v1og6oa4bj, plumbanetellone, bleitellurid, lead monotelluride, lead ii telluride trace metals basis | |
tellanylidenelead |
Specifications
917°C | |
Pieces | |
MFCD00016274 | |
14,5422 | |
Insoluble in water. | |
[Te]=[Pb] | |
334.80 | |
334.79 | |
Product specific | |
(metals basis) |
1314-91-6 | |
PbTe | |
UN3284 | |
lead telluride, lead ii telluride, unii-v1og6oa4bj, v1og6oa4bj, plumbanetellone, bleitellurid, lead monotelluride, lead ii telluride trace metals basis | |
OCGWQDWYSQAFTO-UHFFFAOYSA-N | |
tellanylidenelead | |
4389803 | |
99.999% | |
8.164 g/mL | |
Lead (II) telluride |
Safety and Handling
GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H302+H332-H350-H360Df-H373
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 215-247-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Lead(II) telluride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.