missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) tetrafluoroborate, 50% w/w aq. soln., Thermo Scientific Chemicals

Product Code. 11334947
Click to view available options
Quantity:
1 kg
3 x 1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in Tin-Lead Electroplating

Used in tin-lead electroplatingLead(II) tetrafluoroborate is used as a catalyst involved in calixarene formation through hydrogen-bonded dimmers and in the preparation of linear polyesters. It is involved in the development and optimization of a bipolar lead-boron tetrafluoride accumulator. It is an electroplating solution for coating metal objects with lead and curing agent for epoxy resins. Further, it is used in printing lines, tin-lead alloy plating, lead soldering and also as a circuit board tin-lead alloy plating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13814-96-5
Molecular Formula B2F8Pb
Molecular Weight (g/mol) 380.80
MDL Number MFCD00016270
InChI Key UQGXAENNKXXYNF-UHFFFAOYSA-N
Synonym lead ii tetrafluoroborate, lead fluoroborate, lead ii tetrafluoroborate solution, lead boron fluoride, lead tetrafluoroborate, unii-wax1e0vj84, lead ii fluoroborate, acmc-1bpsn, wax1e0vj84, lead 2+ ditetrafluoroborate
PubChem CID 12598180
SMILES [Pb++].F[B-](F)(F)F.F[B-](F)(F)F

Specifications

Color Colorless
Physical Form Liquid
Quantity 1 kg
Solubility Information Miscible with water.
IUPAC Name lead(2+);ditetrafluoroborate
Formula Weight 380.81
Odor Odorless
Density 1.62
Chemical Name or Material Lead(II) tetrafluoroborate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) tetrafluoroborate, 50% w/w aqueous solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Lead(II) tetrafluoroborate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'