missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) zirconium oxide, 99.7% (metals basis excluding Hf), Hf 1% max, Thermo Scientific Chemicals

Product Code. 11366988
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

It is practically applied in the area of electroceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12060-01-4
Molecular Formula O3PbZr
Molecular Weight (g/mol) 346.421
MDL Number MFCD00016275
InChI Key RSCSNKLWUCUPDE-UHFFFAOYSA-N
Synonym lead zirconate, o3zr.pb, dioxido oxo zirconium;lead 2+, $l^ 2-lead 2+ ion oxozirconiumbis olate
PubChem CID 16212542
IUPAC Name dioxido(oxo)zirconium;lead(2+)
SMILES [O-][Zr](=O)[O-].[Pb+2]

Specifications

Physical Form Powder
Quantity 500 g
UN Number UN3077
Solubility Information Insoluble in all solvents.
Formula Weight 346.42
Percent Purity 99.7%
Odor Odorless
Density 7.0 g/mL
Assay (metals basis excluding Hf), Hf 1% max.
Chemical Name or Material Lead(II) zirconium oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'