missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) zirconium oxide, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Product Code. 11385047
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12060-01-4
Molecular Formula O3PbZr
Molecular Weight (g/mol) 346.421
MDL Number MFCD00016275
InChI Key RSCSNKLWUCUPDE-UHFFFAOYSA-N
Synonym lead zirconate, o3zr.pb, dioxido oxo zirconium;lead 2+, $l^ 2-lead 2+ ion oxozirconiumbis olate
PubChem CID 16212542
IUPAC Name dioxido(oxo)zirconium;lead(2+)
SMILES [O-][Zr](=O)[O-].[Pb+2]

Specifications

Physical Form Powder
Quantity 10 g
UN Number UN3077
Formula Weight 346.42
Percent Purity 99.997%
Grade Puratronic™
Odor Odorless
Density 7.0 g/mL
Assay (metals basis)
Chemical Name or Material Lead(II) zirconium oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'