Learn More
Lead in Isooctane standard solution, Specpure™, 3.70μg/g(0.010g/gal)
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS | 540-84-1 |
Quantity | 100 g |
Odor | Nearly odorless |
Molecular Formula | C8H18 |
MDL Number | MFCD01094200 |
UN Number | UN1262 |
Physical Form | Liquid |
Color | Colorless |
InChI Key | NHTMVDHEPJAVLT-UHFFFAOYSA-N |
SMILES | CC(C)CC(C)(C)C |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1493502 | Certificate of Analysis | 15/10/2024 | 045333.22 |
1453634 | Certificate of Analysis | 13/05/2024 | 045333.22 |
1390146 | Certificate of Analysis | 24/10/2023 | 045333.22 |
1265001 | Certificate of Analysis | 09/05/2023 | 045333.22 |
1266283 | Certificate of Analysis | 08/11/2022 | 045333.22 |
Safety and Handling



- Lead in Isooctane standard solution, Specpure, 3.70µg/g (0.010g/gal)
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Aspiration toxicity Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H304-May be fatal if swallowed and enters airways.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : Isooctane
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.