missing translation for 'onlineSavingsMsg'
Learn More

Lead(IV) oxide, 97+%, extra pure, Thermo Scientific Chemicals

46.53 € - 711.54 €

Chemical Identifiers

CAS 1309-60-0
Molecular Formula O2Pb
Molecular Weight (g/mol) 239.20
MDL Number MFCD00011165
InChI Key YADSGOSSYOOKMP-UHFFFAOYSA-N
Synonym lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french
PubChem CID 14793
SMILES O=[Pb]=O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10246320
SDS View Documents
Thermo Scientific Acros
217530050
5 g Glass bottle
46.53 €
5g
Please to purchase this item. Need a web account? Register with us today!
10092960
SDS View Documents
Thermo Scientific Acros
217531000
100 g Glass bottle
54.17 €
100g
Please to purchase this item. Need a web account? Register with us today!
10480681
SDS View Documents
Thermo Scientific Acros
217535000
500 g Glass bottle
182.91 €
500g
Please to purchase this item. Need a web account? Register with us today!
10350544
SDS View Documents
Thermo Scientific Acros
217530025
2.5 kg Glass Bottle
711.54 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

1309-60-0
239.20
YADSGOSSYOOKMP-UHFFFAOYSA-N
14793
O2Pb
MFCD00011165
lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french
O=[Pb]=O

Specifications

290°C
Brown to Black
97+%
PbO2
01,5331; 02,233; 03,168; 11,282
lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french
YADSGOSSYOOKMP-UHFFFAOYSA-N
dioxolead
14793
≥97%
Lead(IV) oxide, p.a.
1309-60-0
Powder
O2Pb
MFCD00011165
15, 5462
Solubility in water: insoluble. Other solubilities: soluble in hcl and glacial acetic acid,insoluble in alcohol
O=[Pb]=O
239.20
239.19
Extra Pure
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
May damage the unborn child.
Suspected of damaging fertility.

GHS P Statement
Use personal protective equipment as required.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON C

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 215-174-5

missing translation for 'rtecsNumber' : OG0700000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.