Learn More
Lead lanthanum zirconium titanium oxide, polymeric precursor, Thermo Scientific™
Piezoelectric and electro-optic devices
29.30 € - 362.00 €
Specifications
Chemical Name or Material | Lead lanthanum zirconium titanium oxide |
---|---|
Name Note | Polymeric precursor |
Color | Brown |
Physical Form | Viscous Liquid |
MDL Number | MFCD02684260 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Lead lanthanum zirconium titanium oxide | |
Brown | |
MFCD02684260 | |
Soluble in hexane, toluene and alcohol |
Polymeric precursor | |
Viscous Liquid | |
UN1993 | |
Moisture sensitive |
Safety and Handling
GHS H Statement
H360-H373-H226-H302-H332-H336
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P308+P313-P330-P370+P378q-P501c
H226-H302+H332-H336-H360Df-H373
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Lead lanthanum zirconium titanium oxide, polymeric precursor
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P201-Obtain special instructions before use.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P501-Dispose of contents/container to .
Your input is important to us. Please complete this form to provide feedback related to the content on this product.