missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead Perchlorate solution EU Pharmacopoeia 0.1M, Fisher Chemical™
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
Specifications
Specifications
CAS | 33820-53-0 |
Quantity | 1000 mL |
Molecular Formula | C15H23N3O4 |
Synonym | isopropalin, isopropaline, paarlan, 4-isopropyl-2,6-dinitro-n,n-dipropylaniline, caswell no. 506a, isopropaline iso-french, unii-52gzd1i41n, benzenamine, 4-1-methylethyl-2,6-dinitro-n,n-dipropyl, cumidine, 2,6-dinitro-n,n-dipropyl, isopropalin ansi:bsi:iso |
InChI Key | NEKOXWSIMFDGMA-UHFFFAOYSA-N |
SMILES | CCCN(CCC)C1=C(C=C(C=C1[N+](=O)[O-])C(C)C)[N+](=O)[O-] |
IUPAC Name | 2,6-dinitro-4-propan-2-yl-N,N-dipropylaniline |
Molecular Weight (g/mol) | 309.366 |
PubChem CID | 36606 |
ChEBI | CHEBI:82189 |
Show More |
Safety and Handling
Product Identifier
- Lead perchlorate 0.1M Volumetric Solution (RP 311)
- Danger
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360-May damage fertility or the unborn child.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P330-Rinse mouth.
- P391-Collect spillage.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction