missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead shot, 4mm (0.2in), 97% (metals basis), Thermo Scientific Chemicals
36.58 € - 240.20 €
Chemical Identifiers
CAS | 7439-92-1 |
---|---|
Molecular Formula | Pb |
Molecular Weight (g/mol) | 207.20 |
MDL Number | MFCD00134050 |
InChI Key | WABPQHHGFIMREM-UHFFFAOYSA-N |
Synonym | plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant |
PubChem CID | 5352425 |
ChEBI | CHEBI:27889 |
IUPAC Name | lead |
SMILES | [Pb] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15480967
|
Thermo Scientific Alfa Aesar
000675.22 |
100 g |
36.58 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11346617
|
Thermo Scientific Alfa Aesar
000675.36 |
500 g |
83.62 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11356617
|
Thermo Scientific Alfa Aesar
000675.A3 |
2 kg |
240.20 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-92-1 | |
207.20 | |
WABPQHHGFIMREM-UHFFFAOYSA-N | |
5352425 | |
lead |
Pb | |
MFCD00134050 | |
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant | |
CHEBI:27889 | |
[Pb] |
Specifications
7439-92-1 | |
Pb | |
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant | |
[Pb] | |
207.20 | |
CHEBI:27889 | |
Shot |
97% (metals basis) | |
MFCD00134050 | |
WABPQHHGFIMREM-UHFFFAOYSA-N | |
lead | |
5352425 | |
207.2 | |
Lead shot, 4mm (0.2 in.) |
Safety and Handling
P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c
H350-H360FD-H362-H372
missing translation for 'einecsNumber' : 231-100-4
missing translation for 'rtecsNumber' : OF7525000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Lead shot
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only