missing translation for 'onlineSavingsMsg'
Learn More

Lead standard solution, for AAS, 1mg/mL Pb in 2% HNO3, Thermo Scientific Chemicals

23.82 € - 171.86 €

Specifications

Chemical Name or Material Lead standard solution
Name Note 1 mg/ml Pb in 2% HNO3
CAS 10099-74-8, 7697-37-2, 7732-18-5
Packaging Plastic bottle
Density 1.0200g/mL
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10042460
View Documents
Thermo Scientific Acros
196071000
100 mL
23.82 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10052460
View Documents
Thermo Scientific Acros
196075000
500 mL
59.60 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10369133
View Documents
Thermo Scientific Acros
196070025
2.5 L
171.86 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
Specifications

Specifications

Lead standard solution
10099-74-8, 7697-37-2, 7732-18-5
1.0200g/mL
N2O6Pb
Powder/Crystals
15, 5469
RLJMLMKIBZAXJO-UHFFFAOYSA-N
101.0°C
λ²-lead(2+) dinitrate
24924
331.21
1 mg/ml Pb in 2% HNO3
Plastic bottle
1.02
MFCD00011153
997 to 1003 μg/mL of Pb(2+) in 2 to 5% HNO3 (20°C)
Brown to Red or Green
-3.0°C
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
331.20
CHEBI:37187
AAS
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead standard solution, for AAS, 1mg/mL Pb in 2% HNO3, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.