missing translation for 'onlineSavingsMsg'
Learn More

Lead Tin, solder alloy, 1.6mm (0.06in) dia, with rosin core, Thermo Scientific Chemicals

76.88 € - 113.57 €

Specifications

Chemical Name or Material Lead Tin, Solder Alloy
Name Note 1.6mm (0.06 in.) dia., with rosin core
Avg. Mol. Wt. or Mol. Wt. Range Pb:Sn; 50:50 wt%
MDL Number MFCD00798549
Form Solid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11379427
View Documents Promotion Details
Thermo Scientific Alfa Aesar
041024.TB
113 g
76.88 €
113g
Please to purchase this item. Need a web account? Register with us today!
11389427
View Documents Promotion Details
Thermo Scientific Alfa Aesar
041024.29
225 g
113.57 €
225g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lead Tin, Solder Alloy
Pb:Sn; 50:50 wt%
Solid
1.6mm (0.06 in.) dia., with rosin core
MFCD00798549
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c

H302+H332-H350-H360Df-H373

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only