missing translation for 'onlineSavingsMsg'
Learn More

Lead wire, 1.0mm (0.04in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Form: Approx.8.9g/m

167.84 € - 724.61 €

Chemical Identifiers

CAS 7439-92-1
Molecular Formula Pb
Molecular Weight (g/mol) 207.20
MDL Number MFCD00134050
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
Synonym plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
PubChem CID 5352425
ChEBI CHEBI:27889
IUPAC Name lead
SMILES [Pb]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11302008
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011901.G1
1 m
167.84 €
1m
Please to purchase this item. Need a web account? Register with us today!
11391998
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011901.G5
5 m
724.61 €
5m
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7439-92-1
207.20
WABPQHHGFIMREM-UHFFFAOYSA-N
5352425
lead
Pb
MFCD00134050
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
CHEBI:27889
[Pb]

Specifications

Gray
Gray wire
99.998% (Metals basis)
MFCD00134050
WABPQHHGFIMREM-UHFFFAOYSA-N
lead
5352425
Odorless
Lead
7439-92-1
11.34g/cm3 at 20°C
Pb
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
[Pb]
207.20
CHEBI:27889
Wire
Videos
Safety and Handling

Safety and Handling

Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c

H350-H360FD-H362-H372

missing translation for 'einecsNumber' : 231-100-4

missing translation for 'rtecsNumber' : OF7525000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Lead wire

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only