Learn More
Abeba™ Leather Working Shoes
Animal fats, oil and fuel resistant. Sanitized™ treated.
50.05 € - 59.45 €
Specifications
Color Black
Material Leather
View More Specs
Products 8
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15376646
View Documents
Abeba™
7031/35
35 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15396646
View Documents
Abeba™
7031/37
37
56.85 €
1 pair
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
15306656
View Documents
Abeba™
7031/38
38
59.45 €
1 pair
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
15326656
View Documents
Abeba™
7031/40
40
57.75 €
1 pair
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
15336656
View Documents
Abeba™
7031/41
41
56.85 €
1 pair
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
15376656
View Documents
Abeba™
7031/45
45
56.85 €
1 pair
Estimated Shipment: 17-12-2024
to see stock.
Add to basket
15386656
View Documents
Abeba™
7031/46
46
56.85 €
1 pair
Estimated Shipment: 17-12-2024
to see stock.
Add to basket
15396656
View Documents
Abeba™
7031/47
47
56.85 €
1 pair
Estimated Shipment: 17-12-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB FO SRC
  • Smooth leather
  • Black
  • Slip resistance outsole sole
  • Sanitized treated
  • Heel strap foldable, adjustable
  • Leather insole
Specifications
Specifications
Black
Leather