missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Lecithin
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
missing translation for 'unitSize'
500g
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- CAS Number: 8002-43-5
- Soluble in: Chloroform
- Store at: -20°C
Specifications
Specifications
| CAS | 8002-43-5 |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction