missing translation for 'onlineSavingsMsg'
Learn More

Lecithin, MedChemExpress

MedChemExpress Lecithin is regarded as a safe, conventional phospholipid source. Phospholipids are reported to alter the fatty acid composition and microstructure of the membranes in animal cells.

70.20 € - 83.70 €

Specifications

Chemical Name or Material Lecithin
CAS 8002-43-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C42H80NO8P
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16492306
Encompass_Preferred
View Documents
Medchem Express
HY-B2235
500 mg
83.70 €
500mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16468876
Encompass_Preferred
View Documents
Medchem Express
HY-B2235
100 mg
70.20 €
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Lecithin is regarded as a safe, conventional phospholipid source. Phospholipids are reported to alter the fatty acid composition and microstructure of the membranes in animal cells.
Specifications

Specifications

Lecithin
Light Yellow
C42H80NO8P
O=P(OC[C@@H](COC(CCCCCCCCCCCCCCC)=O)OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)(OCC[N+](C)(C)C)[O-]
758.06
Research
8002-43-5
Research
DMSO : 5 mg/mL (6.60 mM; Need ultrasonic) ∣H2O : 3.33 mg/mL (4.39 mM; Need ultrasonic)
758.06
98.0%
Solid
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.