Learn More
Thermo Scientific™ Legionella BCYE Medium
Isolate Legionella from clinical specimens with a non-selective microbiological medium which yields optimal recovery of Legionellaceae in short incubation periods.
Brand: Thermo Scientific™ PO5072A
36.28 EUR valid until 2025-12-31
Use promo code "24364" to get your promotional price.
Description
Isolate Legionella from clinical specimens using Thermo Scientific™ Legionella BCYEα Medium, a non-selective medium that exhibits optimal recovery of legionellaceae in short incubation periods from both environmental samples and clinical specimens.
Legionella are aerobic, motile, nutritionally fastidious, pleomorphic, gram-negative rods that require special growth factors in culture media to grow. Transmission occurs through aerosolization or aspiration of contaminated water, not person to person. Legionellosis refers to two distinct clinical syndromes, Legionnaires’ disease, which most often presents as severe pneumonia, and Pontiac fever, which is an acute, febrile, self-limiting, viral-like illness. Over forty species of Legionella have been characterized, however Legionella pneumophila remains the most prevalent etiological agent of Legionnaires’ disease. Legionella can be isolated from respiratory tract secretions of all types, sterile body fluids such as blood, and biopsy material.Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
Specifications
According to ISO 11731-2 | |
6°C to 12°C | |
90mm Monoplate | |
Agar | |
Flow Wrapped - Single Layer |
Traffic black to Jet black, opaque | |
Legionella BCYEa Medium | |
Germany | |
17g ± 5% | |
10/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.