missing translation for 'onlineSavingsMsg'
Learn More

Novacyt - Microgen Bioproducts Legionella Species latex Reagent

Microgen Bioproducts Legionella Species latex reagent

Brand:  Novacyt - Microgen Bioproducts M45D

Product Code. 17333201

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Individual L.pneumophila serogroups 1, 2-15, and non L.pneumophila species latex and positive control available separately.
Specifications

Specifications

ISO13485 ISO9001
Yes
Legionella species Antibodies
Bacterial Colonies
Reagent
0.85% Isotonic Saline, Disposable Agglutination Slides, Disposable Mixing Sticks, Bacteriological Loops, Legionella Selective Medium, Glass Tubes for Boiling
Polyethylene Dropper Bottle
Monera
2.5 mL
Legionella pneumophila
Bacteria
2°C to 8°C
Non-waived
1 Bottle (2.5 mL)
1
The Confirmatory Identification of Legionella Pneumophila and Other Commonly Isolated Legionellasp Ecies Grown on Selective Medi
2.5 mL
1/Test
Bottle in polygrip bag
M45d Legionella Test Reagent Species 2.5 mL
1
7 minutes
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Product Certifications
Ofertas especiales

Ofertas especiales