missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Leibovitz's L-15 Medium
GREENER_CHOICE

Leibovitz's L-15 Medium

27.32 € - 248.00 €

Specifications

Certifications/Compliance ISO 13485 standards, FDA
Content And Storage 2 to 8°C
Description Leibovitz's L-15 Medium, 500mL, 1 X Concentration, Liquid Form, With Galactose, Phenol Red, L-glutamine, Sodium Pyruvate, 7.4 to 7.9 pH, 12 Months Shelf Life, 300 to 340mOsm/kg Osmolality, Used for Mammalian Cell Culture
Concentration 1 X
Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11570396
GREENER_CHOICE
View Documents
Gibco™
11415049
500 mL
27.32 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11580396
GREENER_CHOICE
View Documents
Gibco™
11415056
10 x 500 mL
248.00 €
5000mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Leibovitz's L-15 Medium supports the growth of HEP-2 monkey kidney cells and primary explants of embryonic and adult human tissue. L-15 is buffered by phosphates and free base amino acids instead of sodium bicarbonate. This medium is designed for supporting cell growth in environments without CO2 equilibration.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ L-15 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ L-15 product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

L-15 contains no proteins, lipids, or growth factors. Therefore, L-15 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS).
Specifications

Specifications

ISO 13485 standards, FDA
Leibovitz's L-15 Medium, 500mL, 1 X Concentration, Liquid Form, With Galactose, Phenol Red, L-glutamine, Sodium Pyruvate, 7.4 to 7.9 pH, 12 Months Shelf Life, 300 to 340mOsm/kg Osmolality, Used for Mammalian Cell Culture
Liquid
Galactose, Phenol Red, Glutamine, Sodium Pyruvate
Mammalian cell culture
For in-vitro diagnostic use
2 to 8°C
1 X
Leibovitz's L-15 Medium
No Glucose, No HEPES, No Sodium Bicarbonate
7.4 to 7.9
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 12 Months

Product Identifier
  • 1,4-Diethylbenzene

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P331-Do NOT induce vomiting.

Supplemental information
  • MIXTURE LIST-Contains : p-Diethyl benzene

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.