Learn More
Gibco™ Leibovitz's L-15 Medium, GlutaMAX™ Supplement
GREENER_CHOICE
Leibovitz's L-15 Medium supports the growth of HEP-2 monkey kidney cells and primary explants of embryonic and adult human tissue. L-15 is buffered by phosphates and free base amino acids instead of sodium bicarbonate.
Brand:  Gibco™ 31415086
 View more versions of this product
Product Code. 11544436
  • 301.00 € / Pack of 10
Explore more special offers
Description
Description
Leibovitz's L-15 Medium supports the growth of HEP-2 monkey kidney cells and primary explants of embryonic and adult human tissue. L-15 is buffered by phosphates and free base amino acids instead of sodium bicarbonate. This medium is designed for supporting cell growth in environments without CO2 equilibration.
This L-15 is modified as follows:
With: Galactose, Phenol Red, GlutaMAX™, Sodium Pyruvate
Without: Glucose, HEPES, Sodium Bicarbonate
The complete formulation is available.
Gibco™ L-15 with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.
L-15 contains no proteins, lipids, or growth factors. Therefore, L-15 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 13485 standards, FDA
Leibovitz's L-15 Medium, 500mL, 1 X Concentration, Liquid Form, With Galactose, Phenol Red, GlutaMAX™, Sodium Pyruvate, 7.4 to 7.9 pH, 12 Months Shelf Life, 300 to 340mOsm/kg Osmolality, Used for Mammalian Cell Culture
Liquid
Sodium Pyruvate
Mammalian cell culture
7.4 to 7.9
For in-vitro diagnostic use
2 to 8°C
1 X
Leibovitz's L-15 Medium
No Glucose, No HEPES, No Sodium Bicarbonate
cGMP-compliant under the ISO 13485 standard
10 x 500 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.