Learn More
Tocris Bioscience™ Lenalidomide 5'-amine
Cereblon ligand with terminal amine for onward chemistry
Brand:  Tocris Bioscience™ 7677/50
277.31 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18738106
  • 351.00 € / 50mg
Explore more special offers
Description
Description
Lenalidomide 5'-amine is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand with a terminal amine for onward chemistry to generate Protein Degraders with rigid linkers. Part of a range of functionalized tool molecules for PROTAC R&D.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
3-(5-Amino-1,3-dihydro-1-oxo-2H-isoindol-2-yl)-2,6-piperidinedione
50 mg
C13H13N3O3
191732-70-4
Degrader Building Blocks