missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Grade 913 Lens Cleaning Tissue
Description
- Very thin paper
- Made with 100% Manilla fibers
- Not fluffy
- Weight: 12g/m2
Application
- Cleaning of various optical lens
- Cleaning of optical fibers filaments
Specifications
Lens Cleaning Tissue | |
Cleaning of various optical lens (objective, microscopes binoculars, magnifying Glass, Glasses, etc.), cleaning of optical fibers filaments | |
500 Sheets |
190 x 135 mm | |
Manilla |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Grade 913 Lens Cleaning Tissue > Dimensions: 190L x 135mmW; Quantity: 500 sheets
Spot an opportunity for improvement?Share a Content Correction