Learn More
Invitrogen™ LentiArray™ Human DNA Damage Response CRISPR Library
The Invitrogen™ LentiArray™ Human DNA Damage Response Library targets 561 genes with up to 4 gRNA per gene target (pooled in a single well) for a total of 2,244 gRNAs. Libraries are delivered as 100 μL of ready to use lentiviral particles per gene target.
Brand:  Invitrogen™ A42281
19980.00 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.
Product Code. 17215996
  • 27000.00 € / Each
Explore more special offers
Description
Show More
Specifications
Specifications
1 library set, 100 μL/target.
Store at -68 to -85°C.
LentiArray™
U6
Dry Ice
Lentiviral
Human CRISPR gRNA Library
1 library
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ LentiArray™ Human DNA Damage Response CRISPR Library > U6