missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Lentil Lectin Sepharose™ 4B Medium

Routinely used in the preparation and purification of a number of glycoproteins and carbohydrate-containing molecules

Brand:  Cytiva 17-0444-01


Product Code. 11544905

  • 862.00 € / 25mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Lentil lectin coupled to Sepharose 4B via cyanogen bromide activation
  • Lentil lectin is a metalloprotein, containing Ca2+ and Mn2+, which binds reversibly to polysaccharides and glycoconjugates containing glucose or mannose type sugars
  • Retains binding in 1% deoxycholate; especially suited for purification of detergent-solubilized membrane proteins
  • Ligand concentration 2 mg Lentil lectin/ml; binding capacity for porcine thyroglobulin approx. 16mg/mL medium
  • Average particle size 90μm
  • Stable in the pH range 3-10, and in the presence of all commonly used buffers (Chelating agents and 8M urea to be avoided)
  • Has more equal affinity for α-D-mannose and α-D-glucose compared to Con A

Order Info

Supplied in 20% ethanol containing 0.9% NaCl, 1mM Ca2+ and 1mM Mn2+

TRUSTED_SUSTAINABILITY
Specifications

Specifications

25mg/mL
4% Agarose
3 to 10
2°C to 8°C
Lentil lectin
2mg Lentil lectin/mL Medium
90 μm
25 mL
Used for purification of a number of Glycoproteins and other Carbohydrate containing molecules
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • lentil lectin sepharose 4B

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers