missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Leuco Crystal Violet, 99%, pure

45.83 € - 141.71 €

Chemical Identifiers

CAS 603-48-5
Molecular Formula C25H31N3
Molecular Weight (g/mol) 373.54
MDL Number MFCD00008314
InChI Key OAZWDJGLIYNYMU-UHFFFAOYSA-N
Synonym 4,4',4''-Methylidynetris(N,N-dimethylaniline), LCV
PubChem CID 69048
IUPAC Name 4-[bis[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)C(C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10138742
View Documents
Thermo Scientific Chemicals
204330050
5 g Glass bottle
45.83 €
5g
Please to purchase this item. Need a web account? Register with us today!
10781091
View Documents
Thermo Scientific Chemicals
204330250
25 g Glass bottle
141.71 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

603-48-5
373.54
OAZWDJGLIYNYMU-UHFFFAOYSA-N
69048
CN(C)C1=CC=C(C=C1)C(C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
C25H31N3
MFCD00008314
4,4',4''-Methylidynetris(N,N-dimethylaniline), LCV
4-[bis[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline

Specifications

175.0°C to 180.0°C
99%
CH[C6H4N(CH3)2]3
13, 315
OAZWDJGLIYNYMU-UHFFFAOYSA-N
CN(C)C1=CC=C(C=C1)C(C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
373.54
373.53
Pure
Powder
603-48-5
C25H31N3
MFCD00008314
4,4',4''-Methylidynetris(N,N-dimethylaniline), LCV
Authentic
4-[bis[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline
69048
99%
Gray to White
Leuco Crystal Violet, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

LIGHT SENSITIVE

missing translation for 'einecsNumber' : 210-043-9

missing translation for 'tsca' : TSCA

Product Identifier
  • Leuco Crystal Violet

Signal Word
  • Warning

Hazard Category
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.