Learn More
3M™ Lexa™ Safety Glasses 15233
Click to view available options
Lens Tint: Clear
Description
- Sleek, curved safety spectacles that are almost invisible when worn
- Rimless and contoured design gives the user an improved field of vision
- Frame is fully customizable with three lens angles and four temple lengths
- DX coating provides enhanced anti-scratch and anti-fog protection
Specifications
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Gray |
Product Type | Safety glasses |
Frame Color | Black |