missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Lexsol Heat Transfer Fluid

Lexsol Heat Transfer Fluid for FreeZone Triad Freeze Dryer or FreeZone Stoppering Tray Dryer.

Brand:  Labconco™ 7840001


Product Code. 17291794

  • 306.00 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For FreeZone Triad Freeze Dryer or FreeZone Stoppering Tray Dryer
Specifications

Specifications

7.57 L
14 in.
13 in.
2 gal.
330.2 mm
355.6 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • LEXSOL SHIPPING KIT 2 GAL

Signal Word
  • Danger

Hazard Category
  • Aspiration toxicity Category 1

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P370+P378-In case of fire: Use water spray, alcohol-resistant foam, drychemical or carbondioxide to extinguish.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : petroleum distilates

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.