missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LH-4000 Fixed Angle Rotor

Work within the parameters of existing protocols with this centrifuge rotor, supporting sample processing applications with capacity ranging from 1.5 to 250mL.

Brand:  Thermo Scientific™ 75006475

3073.25 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


Product Code. 10705424

  • 3296.40 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Work within the parameters of existing protocols with the Thermo Scientific™ LH-4000 Fixed Angle Rotor, supporting a diverse array of sample processing applications. Choose from round or DoubleSpin buckets with a wide array of available adapters providing capacity flexibility ranging from 1.5 to 250mL.

Ordering Information:

Order rotor body (75006476) and components to complete assembly. Tube adapters and microplate adapter for DoubleSpin bucket are also ordered separately. Contact your Sales Representative for information.

Compatible with:

Thermo Scientific Cryofuge 5500i centrifuges.

Note:

Only for use in 230V version

TRUSTED_SUSTAINABILITY
Specifications

Specifications

LH-4000 Fixed Angle Rotor
Swinging Bucket Rotor
5,346 x g
215 mm
Thermo Scientific Sorvall RC 4 Series, Cryofuge 5500i and Multifuge 4 Large Capacity Centrifuges
Round buckets (75006477) and DoubleSpin bucket (75006478) are Biocontainment Certified by CAMR, HPA, Porton Down, U.K.
Yes
Stainless Steel Rotor Body, Aluminum Bucket
4 x 1000 mL
4,400 rpm
88 mm
90°
2,487 x g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.