missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiCl Saturated in Ethanol Electrolyte Solution, SI Analytics™
Click to view available options
Quantity:
250 mL
Description
- Classification according to Regulation (EC) No 1272/2008: GHS02 flame
- Flam. Liq. 2 H225 Highly flammable liquid and vapour
- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources
- Information about fire - and explosion protection: Keep away from sources of ignition and hot surfaces. Protect against electrostatic charges. Wear shoes with conductive soles. Fumes can combine with air to form an explosive mixture. Flammable gas-air mixtures may form in empty receptacles.
- Store away from oxidising agents

Specifications
Specifications
CAS | CAS: 64-17-5, CAS: 7447-41-8 |
Melting Point | −115°C |
Boiling Point | 425°C |
Color | Colorless |
Density | 0.8g/cm3 |
Concentration | 0.1 M |
pH | 7 |
Quantity | 250 mL |
Solution Type | Electrolyte |
Odor | Alcohol-like |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Safety and Handling

Product Identifier
- Ethanol saturated with lithium chloride
- Danger
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- MIXTURE LIST-Contains : ethanol, lithium chloride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LiCl Saturated in Ethanol Electrolyte Solution, SI Analytics™ >
Spot an opportunity for improvement?Share a Content Correction