Learn More
Thermo Scientific™ Lid for Microtiter™ Immulon™ Strip Assemblies
This Thermo Scientific™ Vinyl Lid fits Microtiter Immulon strip plates that contain 1 x 12 strips.
Brand: Thermo Scientific™ 6305
139.78 EUR valid until 2024-10-31
Use promo code "21661" to get your promotional price.
Description
This Thermo Scientific™ Vinyl Lid fits Microtiter Immulon strip plates that contain 1 x 12 strips.
Specifications
Natural | |
Vinyl | |
Cap Strip | |
Microtiter Immulon 1 x 12 Microtiter Strip Assemblies |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.