missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lidocaine, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Anasthetic and class Ib antiarrhythmic agent. Blocks voltage-gated sodium channels in the inactivated state.
Specifications
Specifications
CAS | 137-58-6 |
Quantity | 50 mg |
Molecular Formula | C14H22N2O |
Synonym | lidocaine, lignocaine, xylocaine, lidoderm, 2-diethylamino-n-2,6-dimethylphenyl acetamide, anestacon, esracaine, duncaine, cappicaine, gravocain |
InChI Key | NNJVILVZKWQKPM-UHFFFAOYSA-N |
SMILES | CCN(CC)CC(=O)NC1=C(C=CC=C1C)C |
IUPAC Name | 2-(diethylamino)-N-(2,6-dimethylphenyl)acetamide |
Molecular Weight (g/mol) | 234.343 |
PubChem CID | 3676 |
ChEBI | CHEBI:6456 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'