Learn More
Fisherbrand™ Lids for Desiccator with nz 24/29
For use with desiccators with nz 24/29
Brand:  Fisherbrand™ 632415280202
 View more versions of this product
Product Code. 15524635
  • 172.01 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with desiccators with nz 24/29
Certifications
ISO 13 130, DIN 12 491
Specifications
Specifications
270 mm
Type: 200 mm
97 mm
Lid
Desiccator
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Lids for Desiccator with nz 24/29 > Type: 200mm; Diameter: 270mm; Height: 97mm