Learn More
Fisherbrand™ Lids for Desiccator with nz 24/29
For use with desiccators with nz 24/29
Brand:  Fisherbrand™ 632415280302
 View more versions of this product
Product Code. 15524645
  • 241.00 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with desiccators with nz 24/29
Certifications
ISO 13 130, DIN 12 491
Specifications
Specifications
380 mm
Type: 300 mm
136 mm
Lid
Desiccator