Learn More
Haldenwanger™ Lids for Porcelain Crucibles
Use these lids with porcelain crucibles. Haldenwanger™ Lids for Porcelain Crucibles are constructed from optimized laboratory porcelain.
8.65 € - 98.45 €
Specifications
Material Porcelain
Product Type Specs Crucible Lid
View More Specs
Products 7
Product Code Brand Diameter (Metric) Outer Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer Price Quantity & Availability  
12867713
View Documents Promotion Details
Haldenwanger™
79D/3
68mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12346507
View Documents Promotion Details
Haldenwanger™
79D/9
28mm
67.05 €
Pack of 10
Estimated Shipment: 09-12-2024
to see stock.
Add to basket
12376487
View Documents Promotion Details
Haldenwanger™
79D/8
32mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12356507
View Documents Promotion Details
Haldenwanger™
79D/7
35mm
69.15 €
Pack of 10
Estimated Shipment: 03-12-2024
to see stock.
Add to basket
12306497
View Documents Promotion Details
Haldenwanger™
79D/4
60mm
62.80 €
Pack of 5
Estimated Shipment: 16-12-2024
to see stock.
Add to basket
12366507
View Documents Promotion Details
Haldenwanger™
L 79D/5
52mm
98.45 €
Pack of 10
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
10353121
View Documents Promotion Details
Haldenwanger™
L 79D/5
52mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Available in a variety of sizes
  • For use with Haldenwanger porcelain crucibles
  • Complies with DIN VDE 0335, type C 100
Specifications
Specifications
Porcelain
Crucible Lid