Learn More
Lenz Laborglasintrumente™ Liebig Condenser with Olives
Liebig condenser for condensing vapours
Brand:  Lenz Laborglasintrumente™ 05210201
Product Code. 10200981
  • 56.00 € / Each
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Liebig condenser acc. to DIN 12576 and in addition to DIN
  • With NS socket and NS cone
  • Water connection: glass olives
  • Made of DURAN™ tubing
Specifications
Specifications
DIN 12576
160 mm
NS 14/23
Cooling Surface - 60 cm²
1
DURAN™ Tubing